missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ORAI3 Antibody (S06-8B3), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 421.00 - € 645.00
Specifications
| Antigen | ORAI3 |
|---|---|
| Clone | S06-8B3 |
| Concentration | 0.3 mg/mL |
| Dilution | Western Blot, Immunohistochemistry 1:100, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18649304
|
Novus Biologicals
NBP3-19901-50ul |
50 μg |
€ 421.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18698344
|
Novus Biologicals
NBP3-19901-100ul |
100 μg |
€ 645.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ORAI3 Monoclonal antibody specifically detects ORAI3 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| ORAI3 | |
| 0.3 mg/mL | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Immunology | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 93129 | |
| IgG | |
| Affinity purified |
| S06-8B3 | |
| Western Blot, Immunohistochemistry 1:100, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| ORAI calcium release-activated calcium modulator 3, TMEM142C | |
| Recombinant protein of human Orai3 (Uniprot # Q9BRQ5) | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title