missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ORC3 Polyclonal Antibody

Rabbit Polyclonal Antibody
€ 618.00
Specifications
| Antigen | ORC3 |
|---|---|
| Concentration | 0.05 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17129033
![]() |
Invitrogen™
PA5111111 |
100 μL |
€ 618.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17179293
|
Invitrogen™
PA5111378 |
100 μL |
€ 618.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Immunogen sequence: NNVTPYVVSL QAKDCPDMKH FLQKLISQLM DCCVDIKSKE EESVHVTQRK THYSMDSLSS WYMTVTQKTD PKMLSKKRTT SSQWQSPPVV VILKD Highest antigen sequence indentity to the following orthologs: Rat - 76%, Mouse - 71%.
Histone deacetylase 2 (HDAC2), or transcriptional regulator homolog RPD3 L1, is highly homologous to the yeast transcription factor RPD3 (reduced potassium dependency 3) gene. As in yeast, human HDA2 is likely to be involved in regulating chromatin structure during transcription. It has been implicated to associate with YY1, a mammalian zinc-finger transcription factor, which negatively regulates transcription by tethering RPD3 to DNA as a cofactor. This process is highly concerved from yeast to human.Specifications
| ORC3 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human | |
| ORC3 | |
| homolog of latheo, Drosophila; IMAGE50150; LAT; LATHEO; Orc3; ORC3L; origin of replication 3 homolog; origin recognition complex subunit 3; Origin recognition complex subunit Latheo; origin recognition complex, subunit 3; origin recognition complex, subunit 3 honolog; origin recognition complex, subunit 3-like; OTTHUMP00000040611; RP3-486L4.1 | |
| ORC3 | |
| Primary | |
| Antigen affinity chromatography |
| 0.05 mg/mL | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UBD5 | |
| 23595 | |
| Recombinant Protein corresonding to Human ORC3. Recombinant protein control fragment (Product #RP-94839). | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title