Learn More
Skytec™ Oregon Safety Gloves
Product Code. 30274571
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Unit Size:
10 pairs
Product Code. 30274571
Brand: Skytec™ SKG00002DH
A very thin, flexible and dexterous knuckle coated glove offering outstanding tactility and grip in wet and oily conditions.
  • Ultra Lightweight Glove Nitrile Knuckles Coated and Great Oil Grip
  • Helps to protect the hand from oils
  • Breathable back of hand to reduce perspiration
  • Seamless knit prevents irritation
  • Elasticated wrist provides a secure fit
Specifications
ANSI Abrasion Rating ANSI 3
Certifications/Compliance (Europe) CE II, UKCA, ANSI/ISEA 105-2016: Cut A1,Abrasion 3, EN 388:2016 + A1:2018 4121X
Construction Seamless Knit
Coating Material Nitrile
Color Blue, Black
Exterior Finish Sandpatch
Hand Ambidextrous
Latex Free Yes
Material Nylon
Product Type General Purpose Gloves
Size Large
Liner Material Nylon
Length (Metric) 250 mm