Learn More
Skytec™ Oregon Safety Gloves
Product Code. 30274601
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Unit Size:
10 pairs
Product Code. 30274601
Brand: Skytec™ SKG00002DL
Only null left
Add to basket
Add to basket
A very thin, flexible and dexterous knuckle coated glove offering outstanding tactility and grip in wet and oily conditions.
  • Ultra Lightweight Glove Nitrile Knuckles Coated and Great Oil Grip
  • Helps to protect the hand from oils
  • Breathable back of hand to reduce perspiration
  • Seamless knit prevents irritation
  • Elasticated wrist provides a secure fit
Specifications
ANSI Abrasion Rating ANSI 3
Certifications/Compliance (Europe) CE II, UKCA, ANSI/ISEA 105-2016: Cut A1,Abrasion 3, EN 388:2016 + A1:2018 4121X
Construction Seamless Knit
Coating Material Nitrile
Color Blue, Black
Exterior Finish Sandpatch
Hand Ambidextrous
Latex Free Yes
Material Nylon
Product Type General Purpose Gloves
Size 2X-Large
Liner Material Nylon
Length (Metric) 270 mm