missing translation for 'onlineSavingsMsg'
Learn More

Oxynitrox∣r S100, free radical, 99+%, average M.W. ca 2,250

Product Code. 11411810
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11411810

Brand: Thermo Scientific Alfa Aesar H27530.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 91993-31-6

Steers the selective oxidization of primary alcohols into aldehydes. Additionally, secondary alcohols can be converted into ketones

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91993-31-6
Synonym PIPO

Specifications

Quantity 25 g
Formula Weight 2,250 d (avg.)
Percent Purity ≥99%
Chemical Name or Material Oxynitrox™ S100, free radical

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.