missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p120-catenin Antibody (SR1036), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21614-100ul
This item is not returnable.
View return policy
Description
p120-catenin Monoclonal antibody specifically detects p120-catenin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
Specifications
| p120-catenin | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
| SR1036 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:50-1:100, Immunoprecipitation 1:30 | |
| Cadherin-associated Src substrate, CAS, catenin (cadherin-associated protein), delta 1, catenin delta-1, CTNND, KIAA0384P120CAS, p120, p120 catenin, p120(cas), p120(CTN), p120cas, P120CTN | |
| A synthesized peptide derived from human p120-catenin (Uniprot #: O60716) | |
| 100 μg | |
| Phospho Specific, Signal Transduction | |
| 1500 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction