Learn More
P27 KIP1 Mouse, anti-Human, CoraLite Plus 488, Clone: 3F12C10, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL488-67355-20UL
 View more versions of this product
Product Code. 18018215
  • € 145.00 / 20µL
Description
The P27; KIP1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human P27; KIP1. This antibody recognizes human, mouse, rat antigen. The P27; KIP1 antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
P27 KIP1
3F12C10
CoraLite Plus 488
Mouse
Primary
Human, Mouse, Rat
155 kDa
IgG1
Flow Cytometry (Intracellular Staining)
1000 μg/mL
CDKN1B, CDKN4, KIP1, MEN1B, MEN4, P27, P27KIP1
20 μL
1027
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.