Learn More
P27 KIP1 Mouse, anti-Human, CoraLite Plus 488, Clone: 3F12C10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-67355-20UL
Description
The P27; KIP1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human P27; KIP1. This antibody recognizes human, mouse, rat antigen. The P27; KIP1 antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™ plus 488.
Specifications
| P27 KIP1 | |
| 3F12C10 | |
| CoraLite Plus 488 | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat | |
| 155 kDa | |
| IgG1 |
| Flow Cytometry (Intracellular Staining) | |
| 1000 μg/mL | |
| CDKN1B, CDKN4, KIP1, MEN1B, MEN4, P27, P27KIP1 | |
| 20 μL | |
| 1027 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |