Learn More
P27 KIP1 Mouse, anti-Human, CoraLite Plus 488, Clone: 3F12C10, Proteintech
Mouse Monoclonal Antibody
€ 145.00 - € 450.00
Specifications
| Antigen | P27 KIP1 |
|---|---|
| Clone | 3F12C10 |
| Concentration | 1000 μg/mL |
| Applications | Flow Cytometry (Intracellular Staining) |
| Conjugate | CoraLite Plus 488 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18018215
|
Proteintech
CL488-67355-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18047915
|
Proteintech
CL488-67355-100UL |
100 μL |
€ 450.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The P27; KIP1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human P27; KIP1. This antibody recognizes human, mouse, rat antigen. The P27; KIP1 antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™ plus 488.
Specifications
| P27 KIP1 | |
| 1000 μg/mL | |
| CoraLite Plus 488 | |
| Mouse | |
| 1027 | |
| IgG1 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 3F12C10 | |
| Flow Cytometry (Intracellular Staining) | |
| Liquid | |
| Human, Mouse, Rat | |
| CDKN1B, CDKN4, KIP1, MEN1B, MEN4, P27, P27KIP1 | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.