missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P2X6/P2RX6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-48584-25ul
This item is not returnable.
View return policy
Description
P2X6/P2RX6 Polyclonal antibody specifically detects P2X6/P2RX6 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| P2X6/P2RX6 | |
| Polyclonal | |
| Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| ATP receptor, MGC129625, P2RXL1skeletal muscle-expressed P2X, P2X purinoceptor 6, P2X6purinoceptor P2X6, P2XMpurinoreceptor P2X6, Purinergic receptor, purinergic receptor P2X, ligand-gated ion channel, 6, Purinergic receptor P2X-like 1, purinergic receptor P2X-like 1, orphan receptor | |
| This antibody was developed against a recombinant protein corresponding to amino acids: VTQIKELGNRLWDVADFVKPPQGENVFFLVTNFLVTPAQVQGRCPEHPSVPLAN | |
| 25 μL | |
| Cancer, Cardiovascular Biology, Endocrinology, Neuroscience, Signal Transduction | |
| 9127 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction