missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P2Y12/P2RY12 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NBP1-78249-0.05MG
This item is not returnable.
View return policy
Description
P2Y12/P2RY12 Polyclonal specifically detects P2Y12/P2RY12 in Human, Mouse, Rat, Porcine, Monkey, Primate samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| P2Y12/P2RY12 | |
| Polyclonal | |
| Western Blot 1:500, ELISA 1:10000, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:200, Immunohistochemistry-Paraffin 1:100, Dot Blot 1:10000 | |
| Q9H244 | |
| P2RY12 | |
| Synthetic peptide selected from human P2Y12 receptor gene corresponding to amino acids with the 320-342 at the C-terminus of protein. The peptide was covalently modified to achieve desired antigenicity before coupling to a carrier protein. | |
| Primary | |
| This antibody detects a single 56-68kDa P2Y12 diffused band. The antibody does not cross reacts with other P2X or with other P2Y receptors on western blots. | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Tris/Glycine buffer pH 7.5-7.9, stabilizing protein, and glycerol with 0.02% sodium azide | |
| ADPG-R, Gi-coupled ADP receptor HORK3, HORK3P2Y12 platelet ADP receptor, P2T(AC), P2Y purinoceptor 12, P2Y(AC), P2Y(ADP), P2Y(cyc), P2Y12ADP-glucose receptor, purinergic receptor P2RY12, purinergic receptor P2Y, G-protein coupled, 12, putative G-protein coupled receptor, SP1999G-protein coupled receptor SP1999 | |
| Rabbit | |
| 0.05 mg | |
| 64805 | |
| Human, Rat, Primate | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction