missing translation for 'onlineSavingsMsg'
Learn More

P2Y12/P2RY12 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 3 publications

€ 273.00 - € 726.60

Specifications

Antigen P2Y12/P2RY12
Applications Immunoprecipitation, Immunocytochemistry, Western Blot, ELISA, Immunohistochemistry (Paraffin), Immunofluorescence, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18075676
View Documents
Novus Biologicals
NBP1-78249-0.025MG
0.025 mg
ONLINE EXCLUSIVE PRICE
€ 289.00
€ 273.00 / 25µg
Save € 16.00
5% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18270529
View Documents
Novus Biologicals
NBP1-78249-0.05MG
0.05 mg
ONLINE EXCLUSIVE PRICE
€ 520.00
€ 491.40 / 0.05mg
Save € 28.60
5% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18230116
View Documents
Novus Biologicals
NBP1-78249
0.1 mg
ONLINE EXCLUSIVE PRICE
€ 769.00
€ 726.60 / 0.10mg
Save € 42.40
5% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

P2Y12/P2RY12 Polyclonal specifically detects P2Y12/P2RY12 in Human, Mouse, Rat, Porcine, Monkey, Primate samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications

Specifications

P2Y12/P2RY12
Polyclonal
Rabbit
ADPG-R, Gi-coupled ADP receptor HORK3, HORK3P2Y12 platelet ADP receptor, P2T(AC), P2Y purinoceptor 12, P2Y(AC), P2Y(ADP), P2Y(cyc), P2Y12ADP-glucose receptor, purinergic receptor P2RY12, purinergic receptor P2Y, G-protein coupled, 12, putative G-protein coupled receptor, SP1999G-protein coupled receptor SP1999
P2RY12
Primary
Immunoprecipitation, Immunocytochemistry, Western Blot, ELISA, Immunohistochemistry (Paraffin), Immunofluorescence, Immunohistochemistry
Unconjugated
Q9H244
64805
IgG
This antibody detects a single 56-68kDa P2Y12 diffused band. The antibody does not cross reacts with other P2X or with other P2Y receptors on western blots.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.