missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAK3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 369.00 - € 593.00
Specifications
| Antigen | PAK3 |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18606995
|
Novus Biologicals
NBP2-38890-25ul |
25 μL |
€ 369.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18171328
|
Novus Biologicals
NBP2-38890 |
0.1 mL |
€ 593.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PAK3 Polyclonal specifically detects PAK3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| PAK3 | |
| Polyclonal | |
| Rabbit | |
| Protein Kinase | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| beta-PAK, bPAK, CDKN1A, EC 2.7.11, EC 2.7.11.1, hPAK3, mental retardation, X-linked 47, MRX30, MRX47, Oligophrenin-3, OPHN3, p21 (CDKN1A)-activated kinase 3, p21 protein (Cdc42/Rac)-activated kinase 3, p21-activated kinase 3, PAK-3, PAK3beta, serine/threonine-protein kinase PAK 3 | |
| PAK3 | |
| IgG | |
| Affinity Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| O75914 | |
| 5063 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: DGLDNEEKPPAPPLRMNSNNRDSSALNHSSKPLPMAPEEKNKKARLRSIF | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title