missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PALB2 Antibody (M41-P3F3) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50232-0.025ml
This item is not returnable.
View return policy
Description
PALB2 Monoclonal antibody specifically detects PALB2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Specifications
| PALB2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 | |
| DKFZp667I166, DKFZp686E1054, FANCNFLJ21816, partner and localizer of BRCA2, PNCA3 | |
| Peptide Sequence - KENQFLMPPEE | |
| 0.025 mL | |
| Breast Cancer, Cancer | |
| 79728 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, ELISA, Immunohistochemistry | |
| M41-P3F3 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction