missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PALB2 Antibody (M41-P3F3) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 201.00 - € 368.00
Specifications
| Antigen | PALB2 |
|---|---|
| Clone | M41-P3F3 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 |
| Applications | Western Blot, ELISA, Immunohistochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18674148
|
Novus Biologicals
NBP2-50232-0.025ml |
0.025 mL |
€ 201.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18692478
|
Novus Biologicals
NBP2-50232 |
0.1 mL |
€ 368.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PALB2 Monoclonal antibody specifically detects PALB2 in Human samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
| PALB2 | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Breast Cancer, Cancer | |
| PBS | |
| 79728 | |
| IgG1 κ | |
| Protein A purified |
| M41-P3F3 | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| DKFZp667I166, DKFZp686E1054, FANCNFLJ21816, partner and localizer of BRCA2, PNCA3 | |
| Peptide Sequence - KENQFLMPPEE | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title