missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Palladin Monoclonal Antibody (K1E023_7E3), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA556203
This item is not returnable.
View return policy
Description
Sequence of this protein is as follows: FPKKASRTAR IASDEEIQGT KDAVIQDLER KLRFKEDLLN NGQPRLTYEE RMARRLLGAD SATVFNIQEP EEETANQEYK VSSCEQRLIS EIEYRLERSP VDESGDEVQY
Palladin is a cytoskeletal protein required for organization of normal actin cytoskeleton. Palladin plays roles in establishing cell morphology, motility, cell adhesion and cell-extracellular matrix interactions in a variety of cell types. Palladin may function as a scaffolding molecule with the potential to impact both actin polymerization as well as the assembly of existing actin filaments into higher-order arrays. It binds to proteins that bind to either monomeric or filamentous actin. It localizes at sites where active actin remodeling takes place, such as lamellipodia and membrane ruffles. Palladin's different isoforms may have functional differences. It is also involved in the control of morphological and cytoskeletal changes associated with dendritic cell maturation.
Specifications
| Palladin | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Q8WX93 | |
| PALLD | |
| Recombinant human PALLD (immunogen range is 881-990). | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunoprecipitation, Western Blot | |
| K1E023_7E3 | |
| Unconjugated | |
| PALLD | |
| 2410003B16Rik; 6030492A02; 90kDa actin-associated protein palladin; actin-associated protein palladin; CGI151; CGI-151; FLJ22190; FLJ38193; FLJ39139; involved in cytoskeletal assembly; KIAA0992; MYN; myoneurin; palladin; palladin isoform; palladin pseudogene; palladin, cytoskeletal associated protein; PALLD; PNCA1; Sarcoma antigen NY-SAR-77; SIH002 | |
| Mouse | |
| Protein A | |
| RUO | |
| 23022 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ser du en mulighed for forbedring?Del en indholdskorrektion