missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Palladin Monoclonal Antibody (K1E023_7E3), Invitrogen™
Mouse Monoclonal Antibody
€ 493.00 - € 868.00
Specifications
| Antigen | Palladin |
|---|---|
| Clone | K1E023_7E3 |
| Concentration | 1 mg/mL |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30199115
|
Invitrogen™
MA555823 |
50 μL |
€ 493.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30202641
|
Invitrogen™
MA556203 |
200 μL |
€ 868.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: FPKKASRTAR IASDEEIQGT KDAVIQDLER KLRFKEDLLN NGQPRLTYEE RMARRLLGAD SATVFNIQEP EEETANQEYK VSSCEQRLIS EIEYRLERSP VDESGDEVQY
Palladin Monoclonal antibody specifically detects Palladin in Human samples. It is validated for Immunoprecipitation, Western BlotSpecifications
| Palladin | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| 2410003B16Rik; 6030492A02; 90kDa actin-associated protein palladin; actin-associated protein palladin; CGI151; CGI-151; FLJ22190; FLJ38193; FLJ39139; involved in cytoskeletal assembly; KIAA0992; MYN; myoneurin; palladin; palladin isoform; palladin pseudogene; palladin, cytoskeletal associated protein; PALLD; PNCA1; Sarcoma antigen NY-SAR-77; SIH002 | |
| Recombinant human PALLD (immunogen range is 881-990). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| PALLD |
| K1E023_7E3 | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q8WX93 | |
| 23022 | |
| IgG1 | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title