missing translation for 'onlineSavingsMsg'
Learn More

Palladium(II) nitrate, solution, Pd 10% w/w (cont. Pd)

Product Code. 11470290
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11470290 1 Ea. 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11470290 Supplier Thermo Scientific Alfa Aesar Supplier No. 012621.ND

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10102-05-3 | Pd(NO{3}){2} |

Palladium(II) nitrate is used as a reactant for doping activated carbon for catalysis. It acts as a catalyst for hydrogenation of 1-heptyne and naphthalene. It serves as an important raw material for the preparation of platinum-palladium carbon alloy nanocatalysts for methanol-tolerant oxygen reduction reaction in fuel cells and in the synthesis of copper palladium alloy thin films. Furthermore, it is used in the synthesis of di-phenyl sulfide-modified palladium-titanium dioxide catalysts for acetylene hydrogenation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 10102-05-3
Physical Form Liquid
Quantity 1 Ea.
Molecular Formula Pd(NO{3}){2}
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.