Learn More
Pan-Endothelium Marker, p Ser123, p Ser124 Antibody (HIS52) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-00076-0.025MG
 View more versions of this product
Product Code. 18039126
  • € 187.00 / 25µg
Description
Pan-Endothelium Marker Monoclonal specifically detects Pan-Endothelium Marker in Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Western Blot (Negative).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Pan-Endothelium Marker (p Ser123, p Ser124, p ser123, p ser124)
Monoclonal
0.5 mg/mL
Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1-10 ug/ml, Western Blot (Negative)
Mouse
Affinity Purified
RUO
Rat
IgG1 κ
Immunohistochemistry, Flow Cytometry, Western Blot, Immunohistochemistry (Frozen)
HIS52
Unconjugated
Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide
The immunogen for this antibody is Pan endothelium.
0.025 mg
Primary
Store at 4C. Do not freeze.