missing translation for 'onlineSavingsMsg'
Learn More

Pan-Endothelium Marker, p Ser123, p Ser124 Antibody (HIS52) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

€ 187.00 - € 450.00

Specifications

Antigen Pan-Endothelium Marker (p Ser123, p Ser124, p ser123, p ser124)
Clone HIS52
Concentration 0.5 mg/mL
Dilution Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1-10 ug/ml, Western Blot (Negative)
Applications Immunohistochemistry, Flow Cytometry, Western Blot, Immunohistochemistry (Frozen)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18039126
View Documents
Novus Biologicals
NBP2-00076-0.025MG
0.025 mg
€ 187.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18089326
View Documents
Novus Biologicals
NBP2-00076
0.1 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Pan-Endothelium Marker Monoclonal specifically detects Pan-Endothelium Marker in Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Western Blot (Negative).
Specifications

Specifications

Pan-Endothelium Marker (p Ser123, p Ser124, p ser123, p ser124)
0.5 mg/mL
Immunohistochemistry, Flow Cytometry, Western Blot, Immunohistochemistry (Frozen)
Unconjugated
RUO
Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide
IgG1 κ
Affinity Purified
HIS52
Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1-10 ug/ml, Western Blot (Negative)
Monoclonal
Mouse
Rat
The immunogen for this antibody is Pan endothelium.
Primary
Store at 4C. Do not freeze.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.