missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pancreatic Polypeptide/PP Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 16 publications
Brand: Novus Biologicals NB100-1793-25ug
This item is not returnable.
View return policy
Description
Pancreatic Polypeptide/PP Polyclonal specifically detects Pancreatic Polypeptide/PP in Human, Mouse, Rat, Monkey samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Peptide ELISA.
Specifications
| Pancreatic Polypeptide/PP | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| Obinepitide, pancreatic polypeptide (ppy), pancreatic polypeptide PNPPP, pancreatic polypeptide Y, pancreatic prohormone | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Expected cross-reactivity based on sequence identity: Rat, Dog. | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, ELISA | |
| 0.5 mg/mL | |
| Flow Cytometry, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 3 μg/mL, Immunohistochemistry-Frozen 1:500, Immunoblotting, Peptide ELISA Detection limit 1:32000 | |
| P01298 | |
| PPY | |
| Peptide with sequence C-TRPRYGKRHKEDT corresponding to internal region according to NP_002713.1. | |
| 25 μg | |
| Stem Cell Markers | |
| 5539 | |
| Human, Mouse, Monkey | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction