missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pancreatic Polypeptide/PP Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 16 publications
€ 238.00 - € 603.00
Specifications
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
|---|---|
| Isotype | IgG |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18775963
|
Novus Biologicals
NB100-1793 |
€ 603.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
|
18595990
|
Novus Biologicals
NB100-1793-25ug |
25 μg |
€ 238.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Pancreatic Polypeptide/PP Polyclonal specifically detects Pancreatic Polypeptide/PP in Human, Mouse, Rat, Monkey samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Peptide ELISA.Specifications
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title