Learn More
Pancreatic Polypeptide/PP Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 16 publications
Brand:  Novus Biologicals NB100-1793
 View more versions of this product
Product Code. 18775963
  • € 603.00 / 0.10mg
Description
Pancreatic Polypeptide/PP Polyclonal specifically detects Pancreatic Polypeptide/PP in Human, Mouse, Rat, Monkey samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Peptide ELISA.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Immunohistochemistry, Immunohistochemistry (Paraffin)
Store at -20C. Avoid freeze-thaw cycles.