missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Panendothelial Cell Antigen Antibody (MECA-32), DyLight 755, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NB100-77668IR
This item is not returnable.
View return policy
Description
Panendothelial Cell Antigen Monoclonal specifically detects Panendothelial Cell Antigen in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| Panendothelial Cell Antigen | |
| Monoclonal | |
| DyLight 755 | |
| MECA32, MECA-32, Panendothelial Cell Antigen | |
| Rat | |
| Affinity Purified | |
| RUO | |
| 84094 | |
| Mouse | |
| IgG2a κ |
| Western Blot | |
| MECA-32 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| Plvap | |
| Mouse lymph node stromal cells. | |
| 0.5 mL | |
| Primary | |
| Pan endothelial cells except for CNS & muscle capillaries. | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction