missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Panendothelial Cell Antigen Antibody (MECA-32), DyLight 755, Novus Biologicals™
Rat Monoclonal Antibody
€ 581.00
Specifications
| Antigen | Panendothelial Cell Antigen |
|---|---|
| Clone | MECA-32 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | DyLight 755 |
Description
Panendothelial Cell Antigen Monoclonal specifically detects Panendothelial Cell Antigen in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| Panendothelial Cell Antigen | |
| Western Blot | |
| DyLight 755 | |
| RUO | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| 84094 | |
| Mouse lymph node stromal cells. | |
| Primary | |
| Store at 4°C in the dark. |
| MECA-32 | |
| Monoclonal | |
| Rat | |
| Mouse | |
| MECA32, MECA-32, Panendothelial Cell Antigen | |
| Plvap | |
| IgG2a κ | |
| Affinity Purified | |
| Pan endothelial cells except for CNS & muscle capillaries. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title