missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Parvalbumin Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB120-11427SS
This item is not returnable.
View return policy
Description
Parvalbumin Polyclonal antibody specifically detects Parvalbumin in Human, Mouse, Rat, Feline samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunohistochemistry Free-Floating
Specifications
| Parvalbumin | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| D22S749, MGC116759, parvalbumin, parvalbumin alpha, PV | |
| Parvalbumin Antibody made from purified parvalbumin | |
| 0.025 mg | |
| Neuroscience | |
| 5816 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation | |
| 1.0 mg/mL | |
| Western Blot 0.1 μg/mL, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:100 - 1:200, Immunoprecipitation 5 μg/mL, Immunohistochemistry-Paraffin 1 μg/mL, Immunohistochemistry-Frozen 1:10 - 1:500, Immunohistochemistry Free-Floating reported in scientific literature (PMID 33515957) | |
| P20472 | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Feline | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction