missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Parvalbumin Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 194.00 - € 623.00
Specifications
| Antigen | Parvalbumin |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 0.1 μg/mL, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:100 - 1:200, Immunoprecipitation 5 μg/mL, Immunohistochemistry-Paraffin 1 μg/mL, Immunohistochemistry-Frozen 1:10 - 1:500, Immunohistochemistry Free-Floating reported in scientific literature (PMID 33515957) |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18537283
|
Novus Biologicals
NB120-11427 |
0.1 mg |
€ 623.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18552464
|
Novus Biologicals
NB120-11427SS |
0.025 mg |
€ 194.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Parvalbumin Polyclonal antibody specifically detects Parvalbumin in Human, Mouse, Rat, Feline samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunohistochemistry Free-FloatingSpecifications
| Parvalbumin | |
| Western Blot 0.1 μg/mL, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:100 - 1:200, Immunoprecipitation 5 μg/mL, Immunohistochemistry-Paraffin 1 μg/mL, Immunohistochemistry-Frozen 1:10 - 1:500, Immunohistochemistry Free-Floating reported in scientific literature (PMID 33515957) | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Feline | |
| P20472 | |
| 5816 | |
| IgG | |
| Immunogen affinity purified |
| 1.0 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Neuroscience | |
| PBS | |
| D22S749, MGC116759, parvalbumin, parvalbumin alpha, PV | |
| Parvalbumin Antibody made from purified parvalbumin | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title