missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Parvalbumin Polyclonal Antibody
Chicken Polyclonal Antibody
€ 494.00 - € 1704.00
Specifications
| Antigen | Parvalbumin |
|---|---|
| Concentration | Conc. Not Determined |
| Content And Storage | 4°C |
| Applications | Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17856687
|
Invitrogen™
PA5143545 |
500 μL |
€ 1704.00
500µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17856627
|
Invitrogen™
PA5143579 |
100 μL |
€ 494.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Parvalbumin Polyclonal Antibody for ICC/IF, IHC (P), IHC (PFA), Western Blot
Parvalbumin (PV) is a calcium binding protein expressed in specific muscle fibers and fast-firing neurons. PV consists of a single, unbranched chain of linked amino acids and belongs to a larger group of EF hand proteins. Studies have demonstrated that parvalbumin acts in the decay of calcium in the contraction/ relaxation cycle of fast twitch muscles. This data has shown a positive correlation between the rate of relaxation and the concentration of parvalbumin. Parvalbumin is also expressed in a specific population of GABAergic interneurones which are thought to play a role in maintaining the balance between excitation and inhibition in the cortex as well as the hippocampus. In amyotrophic lateral sclerosis (ALS) patents, parvalbumin immunoreactivity is specifically absent from neuron populations lost early in ALS.Specifications
| Parvalbumin | |
| 4°C | |
| Polyclonal | |
| Liquid | |
| IgY | |
| Human, Mouse, Rat | |
| Pvalb | |
| D22S749; HGNC:9704; MGC116759; PALB1; Parv; parvalbumin; Parvalbumin (calcium binding protein); parvalbumin alpha; PV; Pva; PVALB | |
| Pvalb | |
| Primary |
| Conc. Not Determined | |
| Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Chicken | |
| RUO | |
| PBS with 0.02% sodium azide | |
| P02625, P20472, P32848 | |
| 19293, 25269, 5816 | |
| Full-length recombinant human protein expressed in and purified from E. coli. | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title