missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAWR Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20688-1-AP-20UL
This item is not returnable.
View return policy
Description
The antibody is specific to PAWR.
This gene encodes a tumor suppressor protein that selectively induces apoptosis in cancer cells through intracellular and extracellular mechanisms. The intracellular mechanism involves the inhibition of pro-survival pathways and the activation of Fas-mediated apoptosis, while the extracellular mechanism involves the binding of a secreted form of this protein to glucose regulated protein 78 (GRP78) on the cell surface, which leads to activation of the extrinsic apoptotic pathway. This gene is located on the unstable human chromosomal 12q21 region and is often deleted or mutated different tumors. The encoded protein also plays an important role in the progression of age-related diseases.Specifications
| PAWR | |
| Polyclonal | |
| Unconjugated | |
| PAWR | |
| Par 4, PAR4, PAWR, Prostate apoptosis response 4 protein | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 114774, 5074, 64513 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q62627, Q925B0, Q96IZ0 | |
| Pawr | |
| Peptide | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction