missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pax7 Mouse, Clone: PAX7/497, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 4 publications
Brand: Novus Biologicals NBP2-32894-0.1MG
This item is not returnable.
View return policy
Description
Pax7 Monoclonal specifically detects Pax7 in Human, Mouse, Rat, Chicken, Zebrafish samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.
Specifications
| Pax7 | |
| Monoclonal | |
| Unconjugated | |
| PBS with 0.05% BSA. with 0.05% Sodium Azide | |
| FLJ37460, HuP1, paired box 7, paired box gene 7, paired box homeotic gene 7, paired box protein Pax-7, paired domain gene 7, PAX7 transcriptional factor, PAX7B, RMS2 | |
| Mouse | |
| 57 kDa | |
| 0.1 mg | |
| Apoptosis, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
| 5081 | |
| Human, Mouse, Rat, Chicken, Zebrafish | |
| Purified |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| PAX7/497 | |
| Western Blot 0.5-1ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 0.5-1ug/ml | |
| P23759 | |
| PAX7 | |
| Recombinant fragment (aa300-600) of human PAX7 protein | |
| Protein A purified | |
| RUO | |
| Primary | |
| The Pax gene family of nuclear transcription factors is comprised of nine members that function during embryogenesis to regulate the temporal and position-dependent differentiation of cells. In addition, the family is involved in a variety of signal transduction pathways in the adult organism. Mutations in the Pax family of proteins have been linked to disease and cancer in humans. Pax-7 is a protein specifically expressed in cultured satellite cell-derived myoblasts. In situ hybridization reveals that Pax-7 is also expressed in satellite cells residing in adult muscle. A chromosomal aberration in the gene encoding Pax-7 causes rhabdomyosarcoma 2 (RMS2) (also called alveolar rhabdomyosarcoma). | |
| Store at 4C. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction