missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pax7 Mouse, Clone: PAX7/497, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 4 publications
€ 307.00 - € 517.00
Specifications
| Antigen | Pax7 |
|---|---|
| Clone | PAX7/497 |
| Dilution | Western Blot 0.5-1ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 0.5-1ug/ml |
| Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18028693
|
Novus Biologicals
NBP2-32894-0.1MG |
0.1 mg |
€ 517.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18480232
|
Novus Biologicals
NBP2-32894-0.02mg |
0.02 mg |
€ 307.00
0.02mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Pax7 Monoclonal specifically detects Pax7 in Human, Mouse, Rat, Chicken, Zebrafish samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.Specifications
| Pax7 | |
| Western Blot 0.5-1ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 0.5-1ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Chicken, Zebrafish | |
| P23759 | |
| 5081 | |
| Recombinant fragment (aa300-600) of human PAX7 protein | |
| Primary | |
| Store at 4C. | |
| 57 kDa |
| PAX7/497 | |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
| PBS with 0.05% BSA. with 0.05% Sodium Azide | |
| FLJ37460, HuP1, paired box 7, paired box gene 7, paired box homeotic gene 7, paired box protein Pax-7, paired domain gene 7, PAX7 transcriptional factor, PAX7B, RMS2 | |
| PAX7 | |
| IgG1 κ | |
| Protein A purified | |
| The Pax gene family of nuclear transcription factors is comprised of nine members that function during embryogenesis to regulate the temporal and position-dependent differentiation of cells. In addition, the family is involved in a variety of signal transduction pathways in the adult organism. Mutations in the Pax family of proteins have been linked to disease and cancer in humans. Pax-7 is a protein specifically expressed in cultured satellite cell-derived myoblasts. In situ hybridization reveals that Pax-7 is also expressed in satellite cells residing in adult muscle. A chromosomal aberration in the gene encoding Pax-7 causes rhabdomyosarcoma 2 (RMS2) (also called alveolar rhabdomyosarcoma). |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title