missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PD-L1 Antibody (MM0102-9E5) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12046-0.025MG
This item is not returnable.
View return policy
Description
PD-L1 Monoclonal specifically detects PD-L1 in Human samples. It is validated for Western Blot.
Specifications
| PD-L1/B7-H1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:500-1000 | |
| Q9NZQ7 | |
| CD274 | |
| Human CD274 | |
| 0.025 mg | |
| Primary | |
| Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot | |
| MM0102-9E5 | |
| Unconjugated | |
| A 0.2 um filtered solution in PBS with No Preservative | |
| B7-H, B7-H1, B7H1PDCD1L1, CD274 antigenMGC142294, CD274 molecule, PDCD1LG1MGC142296, PD-L1, PD-L1B7 homolog 1, PDL1PDCD1 ligand 1, programmed cell death 1 ligand 1, Programmed death ligand 1 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 29126 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction