missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PD-L1 Antibody (MM0102-9E5) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 185.00 - € 529.00
Specifications
| Antigen | PD-L1/B7-H1 |
|---|---|
| Clone | MM0102-9E5 |
| Concentration | LYOPH |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18057976
|
Novus Biologicals
NBP2-12046-0.025MG |
0.025 mg |
€ 185.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18072587
|
Novus Biologicals
NBP2-12046 |
0.1 mg |
€ 529.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PD-L1 Monoclonal specifically detects PD-L1 in Human samples. It is validated for Western Blot.Specifications
| PD-L1/B7-H1 | |
| LYOPH | |
| Unconjugated | |
| Mouse | |
| Human | |
| 29126 | |
| IgG1 | |
| Protein G purified |
| MM0102-9E5 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Q9NZQ7 | |
| CD274 | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title