missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PD-L1 Monoclonal Antibody (OTI8C8), TrueMAB™, OriGene
Human Monoclonal Antibody
Brand: Origene Technologies TA813015
This item is not returnable.
View return policy
Description
PD-L1 (Programmed death receptor ligand 1, B7-H1, CD274) is a 40 kDa transmembrane protein and B7 family member. PD-L1 has been shown to negatively regulate T-cell receptor (TCR) signaling. Upon ligating its receptor, PD-L1 has been reported to decrease TCR-mediated proliferation and cytokine production, and PD-L1 expression was found to be abundant on many murine and human cancers and could be further up-regulated upon IFN-gamma stimulation. Thus, PD-L1 might play an important role in tumor immune evasion. Together with B7.1, PD-L1 can inactivate T cells and this relationship is a focal point of cancer therapeutic treatment. At least two isoforms of PDL-1 are known to exist.Specifications
| PD-L1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q9NZQ7 | |
| CD274 | |
| Affinity Chromatography | |
| RUO | |
| 29126 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry | |
| OTI8C8 | |
| Unconjugated | |
| CD274 | |
| B7-H, B7H1, PD-L1, PDCD1L1, PDCD1LG1, PDL1 | |
| Hamster | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction