missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PD-L1 Monoclonal Antibody (OTI8C8), TrueMAB™, OriGene
Human Monoclonal Antibody
€ 242.00 - € 540.87
Specifications
| Antigen | PD-L1 |
|---|---|
| Clone | OTI8C8 |
| Concentration | 1 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17577634
|
Origene Technologies
TA813015S |
30 μL |
€ 242.00
30µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17557634
|
Origene Technologies
TA813015 |
100 μL |
€ 540.87
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PD-L1 Monoclonal antibody specifically detects PD-L1 in Human samples. It is validated for Flow CytometrySpecifications
| PD-L1 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| Human | |
| Q9NZQ7 | |
| IgG1 | |
| -20° C, Avoid Freeze/Thaw Cycles |
| OTI8C8 | |
| Flow Cytometry | |
| Unconjugated | |
| RUO | |
| PBS with no preservative | |
| 29126 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title