missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PD-L1 Monoclonal Antibody (OTI8C8), TrueMAB™, OriGene
Human Monoclonal Antibody
Brand: Origene Technologies TA813015S
This item is not returnable.
View return policy
Description
PD-L1 Monoclonal antibody specifically detects PD-L1 in Human samples. It is validated for Flow CytometrySpecifications
| PD-L1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q9NZQ7 | |
| Human | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry | |
| OTI8C8 | |
| Unconjugated | |
| Cd274 | |
| A530045L16Rik; B7 homolog 1; B7-H; B7h1; B7-H1; Cd274; CD274 antigen; CD274 molecule; PDCD1 ligand 1; Pdcd1l1; Pdcd1lg1; Pdl1; PD-L1; pdl-1; Programmed cell death 1 ligand 1; programmed death ligand 1; RGD1566211; sCD274; soluble CD274; soluble PD L1; sPD L1 | |
| Affinity chromatography | |
| RUO | |
| 29126 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction