missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDE6C Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
€ 149.35 - € 422.30
Specifications
| Antigen | PDE6C |
|---|---|
| Concentration | 0.13 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16876665
|
Proteintech
55183-1-AP-20UL |
20 μL |
€ 149.35
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16866665
|
Proteintech
55183-1-AP-150UL |
150 μL |
€ 422.30
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This antibody is specific to PDE6C.
PDE6C is the alpha-prime subunit of cone phosphodiesterase, which is composed of a homodimer of two alpha-prime subunits and 3 smaller proteins of 11, 13, and 15 kDa.Specifications
| PDE6C | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P51160 | |
| 5146 | |
| Peptide | |
| Primary | |
| -20°C | |
| PDE6C |
| 0.13 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| cGMP phosphodiesterase 6C, PDE6C, PDEA2 | |
| PDE6C | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title