Learn More
Invitrogen™ PDLIM5 Monoclonal Antibody (K56010_2H11), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA556120
Description
Sequence of this protein is as follows: PAPWGFRLQG GKDFNMPLTI SSLKDGGKAA QANVRIGDVV LSIDGINAQG MTHLEAQNKI KGCTGSLNMT LQRASAAPKP EPVPVQKGEP KEVVKPVPIT SPAVSKVTST NNMAYNKAPR PFGSVSSPKV TSIPSPSSAF TPAHATTSSH ASPSPVAAVT PPLFAASGLH ANANLSADQS PSALSAGKTA VNVPRQPTVT SVCSETSQEL AEGQRRGSQG DSKQQNGPPR KHIVERYTEF YHVPTHSDAS KKRLIEDTED WRPRTGTTQS RSFRILAQIT GTEHLKESEA DNTKKANNSQ EPSPQLASSV ASTRSMPESL DSPTSGRPGV
May play an important role in the heart development by scaffolding PKC to the Z-disk region. May play a role in the regulation of cardiomyocyte expansion. Overexpression promotes the development of heart hypertrophy. Contributes to the regulation of dendritic spine morphogenesis in neurons. May restrain postsynaptic growth of excitatory synapses.
Specifications
| PDLIM5 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Q96HC4 | |
| PDLIM5 | |
| Recombinant human PDLIM5 (immunogen range is 11-340). | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| K56010_2H11 | |
| Unconjugated | |
| PDLIM5 | |
| 1110001A05Rik; AI987914; C87059; ENH; Enh1; Enh2; Enh3; Enigma homolog; enigma-like LIM domain protein; enigma-like PDZ and LIM domains protein; L9; LIM; Pdlim5; PDZ and LIM domain 5; PDZ and LIM domain protein 5 | |
| Mouse | |
| Protein A | |
| RUO | |
| 10611 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.