Learn More
Invitrogen™ PDLIM5 Monoclonal Antibody (K56010_2H11), Invitrogen™
Mouse Monoclonal Antibody
€ 450.00 - € 676.00
Specifications
| Antigen | PDLIM5 |
|---|---|
| Clone | K56010_2H11 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30208690
|
Invitrogen™
MA556120 |
200 μL |
€ 676.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30208875
|
Invitrogen™
MA555740 |
50 μL |
€ 450.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: PAPWGFRLQG GKDFNMPLTI SSLKDGGKAA QANVRIGDVV LSIDGINAQG MTHLEAQNKI KGCTGSLNMT LQRASAAPKP EPVPVQKGEP KEVVKPVPIT SPAVSKVTST NNMAYNKAPR PFGSVSSPKV TSIPSPSSAF TPAHATTSSH ASPSPVAAVT PPLFAASGLH ANANLSADQS PSALSAGKTA VNVPRQPTVT SVCSETSQEL AEGQRRGSQG DSKQQNGPPR KHIVERYTEF YHVPTHSDAS KKRLIEDTED WRPRTGTTQS RSFRILAQIT GTEHLKESEA DNTKKANNSQ EPSPQLASSV ASTRSMPESL DSPTSGRPGV
PDLIM5 Monoclonal antibody specifically detects PDLIM5 in Human samples. It is validated for Western BlotSpecifications
| PDLIM5 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| 1110001A05Rik; AI987914; C87059; ENH; Enh1; Enh2; Enh3; Enigma homolog; enigma-like LIM domain protein; enigma-like PDZ and LIM domains protein; L9; LIM; Pdlim5; PDZ and LIM domain 5; PDZ and LIM domain protein 5 | |
| Recombinant human PDLIM5 (immunogen range is 11-340). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| PDLIM5 |
| K56010_2H11 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q96HC4 | |
| 10611 | |
| IgG1 | |
| Protein A | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.