Learn More
Invitrogen™ Pembrolizumab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA558368
 View more versions of this product
Product Code. 30284281
  • € 1204.00 / 1mg
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Pembrolizumab (Keytruda) is a humanized monoclonal antibody that is approved by the U.S. Food and Drug Administration for the treatment of patients with unresectable or metastatic melanoma. Pembrolizumab binds to the human cell surface receptor PD-1 (Programmed Cell Death Protein 1) and blocks its interaction with PD-L1, resulting in the activation of T-cell-mediated immune responses against tumor cells.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Pembrolizumab Humanized
Recombinant Monoclonal
Unconjugated
Keytruda; MK-3475
Protein A
RUO
Human
Antibody
IgG4SP
ELISA, Flow Cytometry
1 mg/mL
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Human
1 mg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized