missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentaerythrityl tetrachloride, 97%, Thermo Scientific™
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Neopentyl bromide and pentaerythrityl tetrachloride were shown here to be non-mutagenic to 7 strains of Salmonella typhimurium.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Neopentyl bromide and pentaerythrityl tetrachloride were shown here to be non-mutagenic to 7 strains of Salmonella typhimurium.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Specifications
| Melting Point | 94°C to 96°C |
| Boiling Point | 110°C (12 mmHg) |
| Quantity | 5 g |
| Beilstein | 1698943 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 209.93 |
| Percent Purity | 97% |
| Chemical Name or Material | Pentaerythrityl tetrachloride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction