missing translation for 'onlineSavingsMsg'
Learn More

Pentaerythrityl tetrachloride, 97%, Thermo Scientific™

Product Code. p-7023197
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11417837

Brand: Thermo Scientific Alfa Aesar A16514.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Neopentyl bromide and pentaerythrityl tetrachloride were shown here to be non-mutagenic to 7 strains of Salmonella typhimurium.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3228-99-7
Molecular Formula C5H8Cl4
Molecular Weight (g/mol) 209.92
MDL Number MFCD00000942
InChI Key KPZGRMZPZLOPBS-UHFFFAOYSA-N
Synonym pentaerythrityl tetrachloride, 1,3-dichloro-2,2-bis chloromethyl propane, tetrachloride, tetrakis chloromethyl methane, pentaerythritol tetrachloride, methane, tetrakis chloromethyl, propane, 1,3-dichloro-2,2-bis chloromethyl, ccris 4944, 2,2-bis chloromethyl-1,3-dichloropropane
PubChem CID 76700
IUPAC Name 1,3-dichloro-2,2-bis(chloromethyl)propane
SMILES ClCC(CCl)(CCl)CCl

Specifications

Melting Point 94°C to 96°C
Boiling Point 110°C (12 mmHg)
Quantity 5 g
Beilstein 1698943
Solubility Information Insoluble in water.
Formula Weight 209.93
Percent Purity 97%
Chemical Name or Material Pentaerythrityl tetrachloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.