Learn More
Honeywell™ PERFECT CUTTING™ Diamond Gloves
Click to view available options
Size:
6
7
8
9
10
11
Unit Size:
1 pair
Description
- White Dyneema™ Diamond/lycra™ knitted glove
- Black polyurethane coating on palm and tips of fingers
- The knitting technique helps to obtain a glove that conforms snugly to the hand and good dexterity
- The glove offers maximum comfort and added sturdiness
- The cool Dyneema™ diamond touch gives an added sensation of comfort
- The tight knit (gauge 13) gives the glove perfect support, limits the penetration of dirt and offers a good touch
- The glove's ventilated back optimises its breathability
- The Dyneema™ diamond/lycra™ thread provides outstanding properties against cuts
- The polyurethane coating at the palm and fingertips provides outstanding abrasion resistance
- Its white color enhances the glove's hygiene and cleanliness
- Its black coating enables work in a slightly soiled environment
Specifications
Specifications
| Certifications/Compliance | EN 420-2003 EEC Basic standards, EN 388-2003 Mechanical hazards |
| Description | 13 gauge |
| Color | White, Gray |
| Material | Polyurethane, Lycra |
| Size | 6 |
| Length (Metric) | 23.5 cm |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur