Learn More
Honeywell™ PERFECT CUTTING™ Diamond Gloves
Product Code. p-7159542
Click to view available options
Size:
6
7
8
9
10
11
Unit Size:
1 pair
Product Code. 15806775
Brand: Honeywell™ 233224506
Only null left
Add to basket
Edge
Add to basket
Honeywell™ PERFECT CUTTING™ Diamond Gloves for handling sharp parts in a dry or greasy environment.
  • White Dyneema™ Diamond/lycra™ knitted glove
  • Black polyurethane coating on palm and tips of fingers
  • The knitting technique helps to obtain a glove that conforms snugly to the hand and good dexterity
  • The glove offers maximum comfort and added sturdiness
  • The cool Dyneema™ diamond touch gives an added sensation of comfort
  • The tight knit (gauge 13) gives the glove perfect support, limits the penetration of dirt and offers a good touch
  • The glove's ventilated back optimises its breathability
  • The Dyneema™ diamond/lycra™ thread provides outstanding properties against cuts
  • The polyurethane coating at the palm and fingertips provides outstanding abrasion resistance
  • Its white color enhances the glove's hygiene and cleanliness
  • Its black coating enables work in a slightly soiled environment
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 420-2003 EEC Basic standards, EN 388-2003 Mechanical hazards
Description 13 gauge
Color White, Gray
Material Polyurethane, Lycra
Size 6
Length (Metric) 23.5 cm