Learn More
Honeywell™ PERFECT POLY™ Black Gloves
Product Code. 15806795
Click to view available options
Size:
6
7
8
9
Unit Size:
1 pair
Product Code. 15806795
Brand: Honeywell™ 240025107
Honeywell™ PERFECT POLY™ Black Gloves for handling fine products in a dry or slightly greasy or soiled environment.
  • Black polyamide knitted glove
  • Grey polyurethane coating on palm and fingertips
  • The knitting technique helps to obtain a glove that conforms snugly to the hand, with good dexterity
  • The glove offers maximum comfort and added sturdiness
  • The tight knit gives the glove perfect support, limits the penetration of dirt and offers a good touch
  • The ventilated back and PU coating optimise the glove's breathability
  • Polyamide with its polyurethane coating provide excellent abrasion and tear resistance
  • The PU coating ensures a good grip
  • Non pilling, the polyamide is suitable for precision work
  • The gloves are certified as silicone-free to ensure their use in painting
  • Hygiene Its dark color makes it more soilproof, thus ensuring a longer life
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 420-03 Basic standard, EN 388-03 Mechanical hazards
Description 13 gauge
Color Black, Gray
Material Polyamide, Polyurethane
Size 7
Length (Metric) 25 cm