Learn More
PERK Antibody (5G5), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand:  Novus Biologicals NBP1-51661-0.025ml
 View more versions of this product
Product Code. 18562284
  • € 205.00 / 25µL
Description
PERK Monoclonal specifically detects PERK in Human samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PERK
Monoclonal
Unconjugated
DKFZp781H1925, EC 2.7.11.1, eukaryotic translation initiation factor 2 alpha kinase 3, eukaryotic translation initiation factor 2-alpha kinase 3, hsPEK, PEKPERKPancreatic eIF2-alpha kinase, PRKR-like endoplasmic reticulum kinase, WRS
Mouse
125 kDa
0.025 mL
Protein Kinase
9451
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
IgG1
Western Blot
5G5
Western Blot 1:500-1:2000, ELISA 1:10000
EIF2AK3
Purified recombinant fragment of human EIF2AK3 expressed in E. Coli.
Unpurified
RUO
Primary
Human
Ascites