missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PERK Antibody (5G5), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
€ 205.00
Specifications
| Antigen | PERK |
|---|---|
| Clone | 5G5 |
| Dilution | Western Blot 1:500-1:2000, ELISA 1:10000 |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
PERK Monoclonal specifically detects PERK in Human samples. It is validated for Western Blot, ELISA.Specifications
| PERK | |
| Western Blot 1:500-1:2000, ELISA 1:10000 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human | |
| 9451 | |
| Purified recombinant fragment of human EIF2AK3 expressed in E. Coli. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| 5G5 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Protein Kinase | |
| DKFZp781H1925, EC 2.7.11.1, eukaryotic translation initiation factor 2 alpha kinase 3, eukaryotic translation initiation factor 2-alpha kinase 3, hsPEK, PEKPERKPancreatic eIF2-alpha kinase, PRKR-like endoplasmic reticulum kinase, WRS | |
| EIF2AK3 | |
| IgG1 | |
| Unpurified | |
| 125 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title