missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGL2 Antibody - BSA Free, Novus Biologicals™
Chicken Polyclonal Antibody
Brand: Novus Biologicals NBP2-41136-0.025mg
This item is not returnable.
View return policy
Description
PGL2 Polyclonal specifically detects PGL2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.
Specifications
| PGL2 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| SDHAF2 | |
| Antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human SDHAF2. The immunogen is located within the first 50 amino acids of SDHAF2. Amino Acid Squence: SFRRFYRGDSPTDSQKD | |
| 0.025 mg | |
| Primary | |
| SDHAF2 antibody is predicted to not cross-react with other SDHAF protein family members. | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
| Western Blot, ELISA | |
| 1 mg/mL | |
| Western Blot 1 μg/mL, ELISA | |
| C11orf79, FLJ20487, hSDH5, paraganglioma or familial glomus tumors 2, PGL2chromosome 11 open reading frame 79, SDH5SDH assembly factor 2, succinate dehydrogenase assembly factor 2, mitochondrial, succinate dehydrogenase complex assembly factor 2, Succinate dehydrogenase subunit 5, mitochondrial | |
| Chicken | |
| Affinity Purified | |
| RUO | |
| 54949 | |
| Human, Mouse, Rat | |
| IgY |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction