missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGL2 Antibody - BSA Free, Novus Biologicals™
Chicken Polyclonal Antibody
€ 168.00 - € 507.00
Specifications
| Antigen | PGL2 |
|---|---|
| Concentration | 1 mg/mL |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18636555
|
Novus Biologicals
NBP2-41136-0.025mg |
0.025 mg |
€ 168.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18116428
|
Novus Biologicals
NBP2-41136 |
0.1 mg |
€ 507.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PGL2 Polyclonal specifically detects PGL2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.Specifications
| PGL2 | |
| Western Blot, ELISA | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| 54949 | |
| Antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human SDHAF2. The immunogen is located within the first 50 amino acids of SDHAF2. Amino Acid Squence: SFRRFYRGDSPTDSQKD | |
| Primary | |
| SDHAF2 antibody is predicted to not cross-react with other SDHAF protein family members. |
| 1 mg/mL | |
| Polyclonal | |
| Chicken | |
| Human, Mouse, Rat | |
| C11orf79, FLJ20487, hSDH5, paraganglioma or familial glomus tumors 2, PGL2chromosome 11 open reading frame 79, SDH5SDH assembly factor 2, succinate dehydrogenase assembly factor 2, mitochondrial, succinate dehydrogenase complex assembly factor 2, Succinate dehydrogenase subunit 5, mitochondrial | |
| SDHAF2 | |
| IgY | |
| Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title