Learn More
PGRMC1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 13 publications
Brand:  Novus Biologicals NBP1-83220
 View more versions of this product
Product Code. 18717173
  • € 518.00 / 0.10mL
Description
PGRMC1 Polyclonal specifically detects PGRMC1 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Western Blot, Immunohistochemistry (Paraffin)
Polyclonal
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.