Learn More
PGRMC1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 13 publications
Brand: Novus Biologicals NBP1-83220
This item is not returnable.
View return policy
Description
PGRMC1 Polyclonal specifically detects PGRMC1 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.
Specifications
| Western Blot, Immunohistochemistry (Paraffin) | |
| Polyclonal | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |