missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PHLPP2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30962-100ug
This item is not returnable.
View return policy
Description
PHLPP2 Polyclonal antibody specifically detects PHLPP2 in Human samples. It is validated for ELISA,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| PHLPP2 | |
| Polyclonal | |
| Western Blot, ELISA, Immunohistochemistry-Paraffin : 10-15 μg/mL | |
| EC 3.1.3.16, KIAA0931PH domain leucine-rich repeat-containing protein phosphatase 2, PH domain and leucine rich repeat protein phosphatase 2, PH domain and leucine rich repeat protein phosphatase-like, PH domain leucine-rich repeat-containing protein phosphatase-like, PHLPPLPHLPP-like | |
| Synthetic peptide derived from the human PHLPP2 protein | |
| 100 μg | |
| Cancer, Tumor Suppressors | |
| 23035 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
| ELISA, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Phosphate buffered saline | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction